AC5H30533DD4 Accela ChemBio Inc.

3,4-Dichloro-5-hydroxy-1-(2-morpholinoethyl)-1H-pyrrol-2(5H)-one

  • MDL number MFCD31716117
  • Cas Number : 1976033-03-0
  • Molecular Weight : 281.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238075&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238075.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart