AC5H30533DD8 Accela ChemBio Inc.

6-Cyclopropylnicotinonitrile

  • MDL number MFCD18255574
  • Cas Number : 1032527-28-8
  • Molecular Weight : 144.17322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238208&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238208.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart