AC5H30533DD9 Accela ChemBio Inc.

Methyl (2S,4S,5R)-3-Boc-2-(4-methoxyphenyl)-4-phenyloxazolidine-5-carboxylate

  • MDL number MFCD31699923
  • Cas Number : 153652-75-6
  • Molecular Weight : 413.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238223&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238223.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart