AC5H30533DDA Accela ChemBio Inc.

3-Methyl-4-quinolinol

  • MDL number MFCD09264039
  • Cas Number : 64965-46-4
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238243&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238243.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart