AC5H30533DDB Accela ChemBio Inc.

1,5-Dihydrobenzo[1,2-d:4,5-d']bis([1,2,3]triazole)

  • MDL number MFCD02684938
  • Cas Number : 7221-63-8
  • Molecular Weight : 160.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238247&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238247.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart