AC5H30533DDD Accela ChemBio Inc.

5-Iodo-4-methyl-2-nitroaniline

  • MDL number MFCD25969656
  • Cas Number : 97113-38-7
  • Molecular Weight : 278.04715

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238258&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238258.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart