AC5H30533DDE Accela ChemBio Inc.

4-Amino-5-methylpyridine-2-methanol

  • MDL number MFCD28724067
  • Cas Number : 1805513-80-7
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238259&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238259.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart