AC5H30533DE1 Accela ChemBio Inc.

4-(4-Ethynylphenyl)morpholine

  • MDL number MFCD00168830
  • Cas Number : 41876-72-6
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY243458&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY243458.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart