AC5H30533DE2 Accela ChemBio Inc.

3-(4-Amino-2-pyridyl)-1-propanol

  • MDL number MFCD18802421
  • Cas Number : 340006-72-6
  • Molecular Weight : 152.19368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246026&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246026.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart