AC5H30533DE4 Accela ChemBio Inc.

1,4-Dioxaspiro[4.4]nonan-7-one

  • MDL number MFCD24705466
  • Cas Number : 109459-59-8
  • Molecular Weight : 142.1525

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246036&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246036.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart