AC5H30533DE5 Accela ChemBio Inc.

tert-Butyl 2-(Diethoxyphosphoryl)-3-methoxypropanoate

  • MDL number MFCD31716134
  • Cas Number : 1286756-55-5
  • Molecular Weight : 296.297061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246051&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246051.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart