AC5H30533DE7 Accela ChemBio Inc.

1-Propyl-8-(4-sulfophenyl)xanthine Potassium Salt Hydrate

  • MDL number MFCD11046025
  • Molecular Weight : 406.4554

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246088&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246088.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart