AC5H30533DEB Accela ChemBio Inc.

Ethyl 3-Amino-4-bromophenylacetate

  • MDL number MFCD18395770
  • Cas Number : 1261677-54-6
  • Molecular Weight : 258.11178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246157&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246157.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart