AC5H30533DED Accela ChemBio Inc.

4,4'-Oxalyldibenzoic Acid

  • MDL number MFCD31716135
  • Cas Number : 1585-67-7
  • Molecular Weight : 298.247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246167&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246167.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart