AC5H30533DEE Accela ChemBio Inc.

5-Methoxy-4-azaindole-2-carbaldehyde

  • MDL number MFCD08448191
  • Cas Number : 17288-50-5
  • Molecular Weight : 176.17202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246189&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246189.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart