AC5H30533DF3 Accela ChemBio Inc.

3-Bromo-2-(difluoromethyl)-6-fluoropyridine

  • MDL number MFCD25477701
  • Cas Number : 1803695-57-9
  • Molecular Weight : 225.9939296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246230&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246230.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart