AC5H30533DF5 Accela ChemBio Inc.

4-(4-Ethynylphenyl)-1-methyl-1H-pyrazole

  • MDL number MFCD30484433
  • Cas Number : 1578111-02-0
  • Molecular Weight : 182.2212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246235&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246235.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart