AC5H30533DF9 Accela ChemBio Inc.

2-Benzyl-5-(trifluoromethyl)pyrazol-3-amine

  • MDL number MFCD11558543
  • Cas Number : 1174306-59-2
  • Molecular Weight : 241.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246244&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246244.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart