AC5H30533DFA Accela ChemBio Inc.

1-(4-Fluoro-3-nitrophenyl)ethanol

  • MDL number MFCD22012648
  • Cas Number : 774-86-7
  • Molecular Weight : 185.1524232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246246&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246246.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart