AC5H30533E00 Accela ChemBio Inc.

1-(2,2-Dimethoxyethyl)-2,4-dinitrobenzene

  • MDL number MFCD31699927
  • Cas Number : 18436-63-0
  • Molecular Weight : 256.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246321&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246321.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart