AC5H30533E01 Accela ChemBio Inc.

3-Methylbenzo[c]isoxazole

  • MDL number MFCD01208009
  • Cas Number : 4127-53-1
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246355&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246355.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart