AC5H30533E04 Accela ChemBio Inc.

2-Bromo-N-methyl-5-(trifluoromethyl)aniline

  • MDL number MFCD29094174
  • Cas Number : 1187395-73-8
  • Molecular Weight : 254.0470896

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246368&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246368.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart