AC5H30533E05 Accela ChemBio Inc.

Ethyl 1-Carbamoylpiperidine-4-carboxylate

  • MDL number MFCD10690931
  • Cas Number : 1092289-81-0
  • Molecular Weight : 200.23494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246369&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246369.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart