AC5H30533E09 Accela ChemBio Inc.

7-Benzyl-2-phenyl-2,7-diazaspiro[4.5]decan-1-one

  • MDL number MFCD13180680
  • Cas Number : 852339-03-8
  • Molecular Weight : 320.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246451&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246451.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart