AC5H30533E0A Accela ChemBio Inc.

6-Chloro-2-(trifluoromethoxy)-3-(trimethylsilyl)pyridine

  • MDL number MFCD22381715
  • Cas Number : 1221172-02-6
  • Molecular Weight : 269.7234496

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246485&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246485.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart