AC5H30533E10 Accela ChemBio Inc.

2-(2-Pyrrolidinyl)aniline

  • MDL number MFCD18705502
  • Cas Number : 1185193-49-0
  • Molecular Weight : 162.23156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246717&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246717.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart