AC5H30533E11 Accela ChemBio Inc.

1-Boc-2-vinyl-4-piperidinone

  • MDL number MFCD30285027
  • Cas Number : 796112-47-5
  • Molecular Weight : 225.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246720&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246720.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart