AC5H30533E12 Accela ChemBio Inc.

3-(2-Allylphenyl)propanoic Acid

  • MDL number MFCD25965854
  • Cas Number : 1352275-74-1
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246721&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246721.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart