AC5H30533E13 Accela ChemBio Inc.

5-Amino-DL-tryptophan

  • MDL number MFCD02683918
  • Cas Number : 6383-69-3
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY246770&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY246770.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart