AC5H30533E20 Accela ChemBio Inc.

5-Bromo-3-chloropyridine-2-carboxamide

  • MDL number MFCD28734539
  • Cas Number : 1335057-63-0
  • Molecular Weight : 235.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247257&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247257.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart