AC5H30533E23 Accela ChemBio Inc.

5-Bromo-N-(3-ethyl-[1,2,4]triazolo[4,3-a]pyridin-8-yl)nicotinamide

  • MDL number MFCD27571324
  • Cas Number : 1340894-13-4
  • Molecular Weight : 346.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247311&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247311.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart