AC5H30533E24 Accela ChemBio Inc.

Diethyl 4-Aminopyridine-2,6-dicarboxylate

  • MDL number MFCD13189673
  • Cas Number : 1196155-10-8
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247366&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247366.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart