AC5H30533E2A Accela ChemBio Inc.

10-Aminoindolo[3,2,1-kl]phenoxazine

  • MDL number MFCD31735641
  • Molecular Weight : 272.30068

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247409&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247409.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart