AC5H30533E2F Accela ChemBio Inc.

4-Fluoro-5-azaindole

  • MDL number MFCD12962624
  • Cas Number : 1190309-76-2
  • Molecular Weight : 136.1264032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247570&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247570.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart