AC5H30533E30 Accela ChemBio Inc.

4,5-Dibromo-3-methylthiophene-2-carbonitrile

  • MDL number MFCD31543713
  • Molecular Weight : 280.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247583&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247583.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart