AC5H30533E31 Accela ChemBio Inc.

6-Methyl-5-nitropyridine-2-methanol

  • MDL number MFCD10699271
  • Cas Number : 13603-40-2
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247595&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247595.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart