AC5H30533E32 Accela ChemBio Inc.

Ethyl 5-Bromo-8-carbamoyl-6-fluoro-2,3,4,9-tetrahydro-1H-carbazole-2-carboxylate

  • MDL number MFCD31735650
  • Cas Number : 1643156-20-0
  • Molecular Weight : 383.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247626&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247626.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart