AC5H30533E34 Accela ChemBio Inc.

2,3,4,6,7,8,9,10-Octahydro-1H-cyclohepta[4,5]pyrrolo[1,2-a]pyrazin-1-one

  • MDL number MFCD31735652
  • Cas Number : 2180068-05-5
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247666&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247666.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart