AC5H30533E36 Accela ChemBio Inc.

3-Ethyl-4-iodo-1H-pyrazole

  • MDL number MFCD07772012
  • Cas Number : 442876-19-9
  • Molecular Weight : 222.02695

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247711&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247711.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart