AC5H30533E3B Accela ChemBio Inc.

5-Amino-4-bromo-1H-indole

  • MDL number MFCD22039260
  • Cas Number : 176713-32-9
  • Molecular Weight : 211.05858

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247754&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247754.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart