AC5H30533E3C Accela ChemBio Inc.

4-(2-Chloroethyl)piperidine Hydrochloride

  • MDL number MFCD12031711
  • Cas Number : 114998-54-8
  • Molecular Weight : 184.1067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247767&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247767.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart