AC5H30533E3D Accela ChemBio Inc.

2-Phenyl-2,7-diazaspiro[4.5]decan-1-one Hydrochloride

  • MDL number MFCD08751989
  • Cas Number : 852338-81-9
  • Molecular Weight : 266.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247771&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247771.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart