AC5H30533E40 Accela ChemBio Inc.

(R)-6-(Boc-amino)-2-hydroxyhexanoic Acid

  • MDL number MFCD31735662
  • Cas Number : 2268819-82-3
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247795&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247795.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart