AC5H30533E44 Accela ChemBio Inc.

Ethyl 7-Bromo-2,4-diethoxy-5H-pyrrolo[3,2-d]pyrimidine-6-carboxylate

  • MDL number MFCD30590390
  • Cas Number : 20419-71-0
  • Molecular Weight : 358.18784

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247870&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247870.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart