AC5H30533E46 Accela ChemBio Inc.

tert-Butyl 2,2-Dichloroacetate

  • MDL number MFCD20621228
  • Cas Number : 49653-47-6
  • Molecular Weight : 185.0484

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247883&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247883.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart