AC5H30533E4D Accela ChemBio Inc.

N-[2-(1-Pyrazolyl)benzyl]-5-(2-hydroxyphenyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31735695
  • Cas Number : 1470368-88-7
  • Molecular Weight : 359.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248124&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248124.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart