AC5H30533E4F Accela ChemBio Inc.

1-Bromo-4-(2-isocyanatoethyl)benzene

  • MDL number MFCD11592307
  • Cas Number : 581812-65-9
  • Molecular Weight : 226.06992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248184&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248184.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart