AC5H30533E51 Accela ChemBio Inc.

5'-Bromospiro[[1,3]dioxolane-2,3'-indolin]-2'-one

  • MDL number MFCD00097597
  • Cas Number : 75822-54-7
  • Molecular Weight : 270.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248189&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248189.pdf
pictograms no data known
Availability details for
Quantity
  • Availability
  • Add to Cart