AC5H3165C43C Accela ChemBio Inc.

(S)-6-[4-(1,1,1-Trifluoro-2-propyl)-4H-1,2,4-triazol-3-yl]-2-pyridinamine

  • MDL number MFCD31811559
  • Molecular Weight : 257.2151096

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY247659&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY247659.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart