AC5H3165C43E Accela ChemBio Inc.

4-(4-Boc-1-piperazinyl)-1-(3-iodophenyl)-2-pyrrolidinone

  • MDL number MFCD31811567
  • Molecular Weight : 471.33251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY248220&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY248220.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart